Learn More
Hach Company Hach Carrier Tray
Supplier:  Hach Company 2938300
Encompass
Steam autoclavable, polypropylene molded tray has built-in handle for easy carrying of reagents or laboratory ware using one hand. Product dimenstions are 14.9 x 9.4 x 4.5" (L x W x H). Each pocket is approximately 4 inches wide.
Catalog No. 50-199-6859