Learn More
Bioworld Carrot Shoot Development Basal Medium, 25 L
Catalog No. 50254891
Encompass_Preferred
Carrot Shoot Development Basal MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required to initiate shoots from carrot callusSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.21 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-891 Supplier Bioworld Supplier No. 306301593
Carrot Shoot Development Basal MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required to initiate shoots from carrot callusSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.21 g/LpH: 3.5–4.5Soluble in water