Learn More
Anton Paar Carrying case for DMA 35 Standard Version or Snap 41
Catalog No. NC1818223
Encompass_Preferred
The plastic carrying case with padding is ideal for secure transportation of your DMA 35 Standard Version 4 or Snap 41 from one sampling location to the next, as well as for storage of your instrument. It has enough space for the instrument (including filling tube) with or without rubber housing as well as additional accessories.Suitable for use with Anton Paar density metersDMA 35 Standard Advanced portable density and concentration meter NC1488853,Snap 41 Portable Alcohol Meter for sugar-free distilled spirits NC1818207
Catalog No. NC1818223 Supplier Anton Paar Supplier No. 177345
The plastic carrying case with padding is ideal for secure transportation of your DMA 35 Standard Version 4 or Snap 41 from one sampling location to the next, as well as for storage of your instrument. It has enough space for the instrument (including filling tube) with or without rubber housing as well as additional accessories.Suitable for use with Anton Paar density metersDMA 35 Standard Advanced portable density and concentration meter NC1488853,Snap 41 Portable Alcohol Meter for sugar-free distilled spirits NC1818207