Learn More
Aviva Systems Biology Corp CARS Antibody - C-terminal region (ARP40295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CARS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CARS. Alternate names: CARS1, CYSRS, MGC:11246ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57379