Learn More
Aviva Systems Biology Corp CARS Antibody - N-terminal region (ARP40297_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40297_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CARS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CARS. Alternate names: CARS1, CYSRS, MGC:11246ApplicationIHC, WB. Species reactivity: Human, Rat
Catalog No. 50-014-57380