Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Optical 4.5X LumiLoupeÖ Power Stand Magnifier
SDPSCI_ED

Catalog No. S13550 Shop All Carson Optical Products
Change view
Click to view available options
Magnification:
4.5x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S13550 4.5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S13550 Supplier Carson Optical Supplier No. LL55
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for viewing insects, plants, maps and graphics

  • 4.5X power magnification
  • Pre-focused dual lenses allows examination of objects close-up
  • Transparent glass base permits ambient light to illuminate the subject
  • Dimensions: 10 x 10 x 7.6cm (4 x 4 x 3 in.)
  • Weight: 280.6g (9.9 oz.)

Specifications

Magnification 4.5x
Product Type Magnifier, Power Stand
For Use With (Application) Magnifier for insects, plants, maps and graphics
Dimensions (L x W x H) 4 x 4 x 3 in.
Weight (English) 9.8 oz.
Lighting Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.