Learn More
Electron Microscopy Sciences Carson's Phosphate Buffered Paraformaldehyde 1 LT
SDP
Catalog No. 501901167
Encompass_Preferred
For some tissues, the use of formalin (contains 10 – 15% Methanol) is not suitable, we offer the alternative formaldehyde (prepared from paraformaldehyde – contains no methanol) for your convenience.
Catalog No. 50-190-1167 Supplier Electron Microscopy Sciences Supplier No. 1573310
For some tissues, the use of formalin (contains 10 – 15% Methanol) is not suitable, we offer the alternative formaldehyde (prepared from paraformaldehyde – contains no methanol) for your convenience.