Learn More
Electron Microscopy Sciences Carson's Phosphate Buffered Formalin 1 LT
SDP
Catalog No. 501902059
Encompass_Preferred
Millonig's phosphate buffered formalin as modified by Carson, Martin and Lynn, by using 4% formalin in 0.1M sodium phosphate monobasic with sodium hydroxide to pH 7.2
Catalog No. 50-190-2059 Supplier Electron Microscopy Sciences Supplier No. 1573210S
Millonig's phosphate buffered formalin as modified by Carson, Martin and Lynn, by using 4% formalin in 0.1M sodium phosphate monobasic with sodium hydroxide to pH 7.2