Learn More
Electron Microscopy Sciences Carson's Phosphate Buffered Formalin 500 ML
SDP
Catalog No. 501901135
Encompass_Preferred
Millonig's phosphate buffered formalin as modified by Carson, Martin and Lynn, by using 4% formalin in 0.1M sodium phosphate monobasic with sodium hydroxide to pH 7.2
Catalog No. 50-190-1135 Supplier Electron Microscopy Sciences Supplier No. 1573205
Millonig's phosphate buffered formalin as modified by Carson, Martin and Lynn, by using 4% formalin in 0.1M sodium phosphate monobasic with sodium hydroxide to pH 7.2