Learn More
Bachem CART (61-102) (rat) Trifluoroacetate
SDP
Supplier:  Bachem 4030510.0100
Encompass_Preferred
CART (61-102) (rat) Trifluoroacetate, H-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH, 209615-75-8, C189H310N58O56S7, Mr 4515.36, 0.1mg. CART (48-89) (human).
Catalog No. 50-259-850