Learn More
BINDER™ Cart with Casters
SDP
Catalog No. 08111310 Shop All BINDER Inc. Products
Change view
Click to view available options
Dimensions (L x W x H):
800 x 1000 x 780 mm
Catalog No. Dimensions (L x W x H)
08-111-310 800 x 1000 x 780 mm
1 options
Catalog No. 08-111-310 Supplier BINDER™ Supplier No. 90510018
Add to Cart
Add to Cart
Stable Cart with Casters with locking brakes. Accessory for use with Avantgarde.Line Incubators.
For Use With:
  • BD 23, BD 56, BD 115, BF 56, BF 115, ED 56, ED 115, FD 56, FD 115, FDL 115, FED 56, FED 115, FP 56, FP 115, KB 53, KB 115, KT 53, KT 115, KT 170, M 53, M 115
Specifications
Description stable cart, casters with locking brakes, dimensions: W 1,000 x D 800 x H 780 mm
For Use With (Equipment) BINDER Chambers BD 23, BD 56, BD 115, BF 56, BF 115, ED 56, ED 115, FD 56, FD 115, FDL 115, FED 56, FED 115, FP 56, FP 115, KB 53, KB 115, KT 53, KT 115, KT 170, M 53, M 115
Material Steel
Type Cart
Dimensions (L x W x H) 800 x 1000 x 780 mm