Learn More
Air Science Usa Base Stand for Ductless Fume Hoods, Laminar Flow Cabinets, PCR Workstations, Vented Enclosures, Downflow Workstations, includes Locking Caster Wheels, 36" / 900mm wide units
SDP
Catalog No. NC0784890
Encompass_Preferred
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-36, DWS36, FLOW36, PCR36, VE34S, Ptefh-36, CA24S, and other 36" / 900mm Nominal Width Models (cart-36)
Catalog No. NC0784890 Supplier Air Science Usa Supplier No. P536CART
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-36, DWS36, FLOW36, PCR36, VE34S, Ptefh-36, CA24S, and other 36" / 900mm Nominal Width Models (cart-36)