Learn More
Chemglass Life Sciences Cart, Heavy Duty, Circulator, Supplied with 10ft Tie Down Strap, For use with Unistat 405 or 705
SDP
Catalog No. 5000980616
Encompass_Preferred
Heavy duty cart for use with Huber Unistat models 405 and 705 circulators. Cart raises the circulators so that the controllers are easily viewed and may make it possible to use shorter hoses. Cart will support up to 500lbs, has a light gray powder coated finish with two locking casters and supplied with one 10ft nylon tie down strap.
Catalog No. 50-009-80616 Supplier Chemglass Life Sciences Supplier No. CG1976H100
Heavy duty cart for use with Huber Unistat models 405 and 705 circulators. Cart raises the circulators so that the controllers are easily viewed and may make it possible to use shorter hoses. Cart will support up to 500lbs, has a light gray powder coated finish with two locking casters and supplied with one 10ft nylon tie down strap.