Learn More
CPC Scientific H-Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt) 0.1MG
SDP
Catalog No. 502108923
Encompass
SEQUENCE: H-Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys20 and 40, Cys14 and 32, Cys34 and 47 bridge)ONE-LETTER SEQUENCE: VPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys20 and 40 bridge, Cys14 and 32 bridge, Cys34 and 47 bridge)MOLECULAR FORMULA: C225H365N65O65S7MOLECULAR WEIGHT: 5245.23STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [214050-22-3]SYNONYMS: CART (55-102) (human)RESEARCH AREA: ObesityREFERENCES:Murphy KG. Brief Funct Genomic Proteomic. 2005, 4(2):95-111.SKU(s): CART-004A, CART-004B
Catalog No. 50-210-8923 Supplier CPC Scientific Supplier No. CART004A
Add to Cart
Add to Cart
SEQUENCE: H-Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys20 and 40, Cys14 and 32, Cys34 and 47 bridge)ONE-LETTER SEQUENCE: VPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys20 and 40 bridge, Cys14 and 32 bridge, Cys34 and 47 bridge)MOLECULAR FORMULA: C225H365N65O65S7MOLECULAR WEIGHT: 5245.23STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [214050-22-3]SYNONYMS: CART (55-102) (human)RESEARCH AREA: ObesityREFERENCES:Murphy KG. Brief Funct Genomic Proteomic. 2005, 4(2):95-111.SKU(s): CART-004A, CART-004B