Learn More
CPC Scientific H-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CART005A
Encompass
SEQUENCE: H-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys13 and 33 bridge, Cys7 and 25 bridge, Cys27 and 40 bridge)ONE-LETTER SEQUENCE: YGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys13 and 33 bridge, Cys7 and 25 bridge, Cys27 and 40 bridge)MOLECULAR FORMULA: C183H298N56O55S7MOLECULAR WEIGHT: 4387.25STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [209615-79-2]SYNONYMS: RESEARCH AREA: ObesityREFERENCES:Thim, Lars, et al. FEBS Letters 428.3 (1998): 263-268.SKU(s): CART-005A, CART-005B
Catalog No. 50-210-8925