Learn More
TrippNT Cart MRI Compact White 1/EA
SDP
Supplier:  TrippNT 54038
Encompass_Preferred
The MRI Compact Cart is purpose-built for MR suites featuring durable HDPE construction and premium all-plastic drawer slides with smooth soft-close detents no metal slides no compromise on quality Its slim footprint fits under counters or in tight spaces offering mobile cleanable and corrosion-resistant storage With 99 4% non-ferrous construction it s safe for MR environments while delivering long-lasting performance and effortless organization where safety and space matter most
Catalog No. 50-005-10087