Learn More
TrippNT Cart MRI Narrow White 1/EA
SDP
Supplier:  TrippNT 51726
Encompass_Preferred
TrippNT s MRI Core SP with White Drawers is compatible for use within the sensitive magnetic imaging environment keeping supplies mobile and organized The cutaway ergonomic grab handles in the top make the Core SP easy to maneuver Constructed from non-ferrous durable high density polyethylene and ABS and non-ferrous casters and screws Ships fully Assembled
Catalog No. 50-005-10085