Learn More
ORS Nasco CART MULTIPURPOSE
Supplier:  ORS Nasco 06875SCL
Encompass_Preferred
Honeywell part# 75SCL. Honeywell North Low-Profile Cartridges and Cartridge/P100 Filter Combinations Cartridge Type: Low Profile Particulate Cartridge Olive Quantity: 18 Case Applications: Industrial applications For Use With: Respirator Defender Multipurpose Cartridge Certifications/Compliance: NIOSH 42CFR84
Catalog No. 50-255-1666