Learn More
Vernier™ Cart Picket Fence
SDPSCI_ED
Catalog No. S15970ND Shop All Vernier Software Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
S15970ND Plastic
1 options
Catalog No. S15970ND Supplier Vernier™ Supplier No. PFCART
Add to Cart
Add to Cart
Includes
Cart Picket Fence
The Cart Picket Fence is a small plastic strip for doing photogate motion timing with dynamics carts. When the Cart Picket Fence passes through a photogate, the cart's velocity and acceleration as a function of time can be accurately measured.
  • The Cart Picket Fence is a small plastic strip for doing photogate motion timing with dynamics carts.
  • When the Cart Picket Fence passes through a photogate, the cart's velocity and acceleration as a function of time can be accurately measured.
Warranty and Services
5-year limited warranty
Specifications
Material Plastic
For Use With (Application) Educational use only
For Use With (Equipment) Photogate, Picket Fence, Dynamics System, Motion Encoder System