Learn More
LabStrong Corp CART PK ORG FREE RO DEION FEED
Catalog No. 501126081
Encompass_Preferred
The LabStrong D50281LS cartridge pack is comparable to the Thermo Scientific™ Barnstead™ D50281 cartridge pack. The LabStrong water purification cartridge pack is to be used with the Thermo Scientific Barnstead NANOpure® DIamond™ water purification systems. This water purification cartridge pack provides outstanding quality you expect from your water system at a low cost without leaks. The cartridge is an economical alternative for all laboratories and easy to install. Plus, all packs include one 0.2μm final filter! LabStrong takes extra care in sealing our water purification cartridges to ensure exceptional quality is maintained throughout the two year shelf life. Product Specifications: • Part Number: D50281LS • Product Type: Water Purification Cartridge Pack • Compatible With: Thermo Scientific Barnstead NANOpure® DIamond™ Style water purification systems • Grain Capacity: 1506 grains • Max Inlet Water Pressure: 100 SI
Catalog No. 50-112-6081 Supplier LabStrong Corp Supplier No. D50281LS
The LabStrong D50281LS cartridge pack is comparable to the Thermo Scientific™ Barnstead™ D50281 cartridge pack. The LabStrong water purification cartridge pack is to be used with the Thermo Scientific Barnstead NANOpure® DIamond™ water purification systems. This water purification cartridge pack provides outstanding quality you expect from your water system at a low cost without leaks. The cartridge is an economical alternative for all laboratories and easy to install. Plus, all packs include one 0.2μm final filter! LabStrong takes extra care in sealing our water purification cartridges to ensure exceptional quality is maintained throughout the two year shelf life. Product Specifications: • Part Number: D50281LS • Product Type: Water Purification Cartridge Pack • Compatible With: Thermo Scientific Barnstead NANOpure® DIamond™ Style water purification systems • Grain Capacity: 1506 grains • Max Inlet Water Pressure: 100 SI