Learn More
CPC Scientific H-Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108919
Encompass
SEQUENCE: H-Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys20 and 40, Cys14 and 32, Cys34 and 47)ONE-LETTER SEQUENCE: IPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys20 and 40 bridge, Cys14 and 32 bridge, Cys34 and 47 bridge)MOLECULAR FORMULA: C226H367N65O65S7MOLECULAR WEIGHT: 5259.34STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [209615-79-2]SYNONYMS: CART (55-102) (rat)RESEARCH AREA: ObesityREFERENCES:SKU(s): CART-002A, CART-002B
Catalog No. 50-210-8919 Supplier CPC Scientific Supplier No. CART002A
SEQUENCE: H-Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys20 and 40, Cys14 and 32, Cys34 and 47)ONE-LETTER SEQUENCE: IPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys20 and 40 bridge, Cys14 and 32 bridge, Cys34 and 47 bridge)MOLECULAR FORMULA: C226H367N65O65S7MOLECULAR WEIGHT: 5259.34STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [209615-79-2]SYNONYMS: CART (55-102) (rat)RESEARCH AREA: ObesityREFERENCES:SKU(s): CART-002A, CART-002B