Learn More
CPC Scientific H-Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108917
Encompass
ONE-LETTER SEQUENCE: IPIYEKKYGQVPMCDAGEQCAV (Cys14 and 20 bridge)MOLECULAR FORMULA: C107H166N26O33S3MOLECULAR WEIGHT:2440.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-15-5], SYNONYMS: CART (55-76) (rat)RESEARCH AREA: Obesity
Catalog No. 50-210-8917 Supplier CPC Scientific Supplier No. CART001A
ONE-LETTER SEQUENCE: IPIYEKKYGQVPMCDAGEQCAV (Cys14 and 20 bridge)MOLECULAR FORMULA: C107H166N26O33S3MOLECULAR WEIGHT:2440.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-15-5], SYNONYMS: CART (55-76) (rat)RESEARCH AREA: Obesity