Learn More
CPC Scientific H-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CART003B
Encompass
ONE-LETTER SEQUENCE: YGQVPMCDAGEQCAV (Cys68 and 74 bridge)MOLECULAR FORMULA: C64H97N17O23S3MOLECULAR WEIGHT:1568.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [210978-19-1], RESEARCH AREA: Obesity
Catalog No. 50-210-8922