Learn More
Cole-Parmer Lakeside Carts 6600 Ergonomic SS cart without guard rails; 24" x 15-1/2", 2 shelves, 300 lb capacity
Catalog No. NC1687032 Shop All Cole-Parmer Products
Encompass_Preferred
Designed to reduce body strain. Vertical push handles keep hands in a comfortable position, reducing stress on arms and wrists. Padding on the handles (all models except 47520-00 and -02) absorbs vibrations and ensures a good grip. The bottom shelf is placed 20" from the floor to prevent potential back strain.
Catalog No. NC1687032 Supplier Cole-Parmer Supplier No. 0094616
Designed to reduce body strain. Vertical push handles keep hands in a comfortable position, reducing stress on arms and wrists. Padding on the handles (all models except 47520-00 and -02) absorbs vibrations and ensures a good grip. The bottom shelf is placed 20" from the floor to prevent potential back strain.