Learn More
VAI Veltek™ Cart2Core™ Utility Cart System Additional Top
Catalog No. 19546589 Shop All VAI Products
Change view
Click to view available options
For Use With (Equipment):
Cart2Core Utility Cart System
Catalog No. For Use With (Equipment)
19-546-589 Cart2Core Utility Cart System
1 options
Catalog No. 19-546-589 Supplier VAI Veltek™ Supplier No. CCUC400T
Add to Cart
Add to Cart
Request a Quote
This is an Additional Top for the Cart2Core™ Utility Cart System.
  • Allows for wheels and bases to be easily and effectively cleaned, disinfected, or sterilized by making the bases and wheels removable
  • Reduces safety concerns with cleaning
  • Provides the ability to steam sterilize bases and wheels routinely
  • Reduces the overuse of disinfectants, therefore, reducing corrosion and pitting
  • Reduces garment contamination and glove ripping
  • Constructed of a sturdy 316L Stainless Steel
  • Fully welded seams that eliminates gaps, unsmooth services, and improves cleaning, disinfection and sterilization
  • Easy pull handle for cart transfer to another base
  • Wheel lock that secures the base during the transfer
  • Easy grip push handles
  • Fits in 4 ft. autoclaves
  • RFID coding for easy location is available/optional
  • 2400 lb. weight capacity (caster rating at 600 psi)
  • 8 point transfer efficiency
Specifications
Product Type Cart Top
For Use With (Equipment) Cart2Core Utility Cart System
Material Stainless Steel