Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI Veltek™ Cart2Core™ Utility Cart System

Catalog No. 19546587 Shop All VAI Products
Change view
Click to view available options
No. of Shelves:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Shelves
19-546-587 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-546-587 Supplier VAI Veltek™ Supplier No. CCUC400
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Cart2Core™ Utility Cart System is an aseptic stainless steel utility cart transfer system that includes a cart top and two bases.

  • Allows for wheels and bases to be easily and effectively cleaned, disinfected, or sterilized by making the bases and wheels removable
  • Reduces safety concerns with cleaning
  • Provides the ability to steam sterilize bases and wheels routinely
  • Reduces the overuse of disinfectants, therefore, reducing corrosion and pitting
  • Reduces garment contamination and glove ripping
  • Constructed of a sturdy 316L Stainless Steel
  • Fully welded seams that eliminates gaps, unsmooth services, and improves cleaning, disinfection and sterilization
  • Easy pull handle for cart transfer to another base
  • Wheel lock that secures the base during the transfer
  • Easy grip push handles
  • Fits in 4 ft. autoclaves
  • RFID coding for easy location is available/optional
  • 2400 lb. weight capacity (caster rating at 600 psi)
  • 8 point transfer efficiency

Specifications

Autoclavable Autoclavable
For Use With (Application) For the transportation of materials between cleanroom classifications that ensures a reduced risk of cross-contamination.
Load Bearing Capacity (English) 2400 lb.
Material Stainless Steel
No. of Shelves 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.