Learn More
Selleck Chemical LLC Carteolol HCl S4278-5mg
SDP
Catalog No. 503109395
Encompass_Preferred
Carteolol HCl(Carteolol hydrochloride OPC-1085 hydrochloride) is a -adrenoceptor antagonist used for the treatment of glaucoma
Catalog No. 50-310-9395 Supplier Selleck Chemical LLC Supplier No. S42785MG
Carteolol HCl(Carteolol hydrochloride OPC-1085 hydrochloride) is a -adrenoceptor antagonist used for the treatment of glaucoma