Learn More
Medchemexpress LLC Carteolol hydrochloride | 51781-21-6 | 99.6% | 328.8343 | 25 MG
SDP
Catalog No. 5000384458
Encompass_Preferred
Carteolol hydrochloride is a non-selective beta blocker and beta-adrenergic antagonist. It is used in research as an anti-arrhythmia agent, an anti-angina agent, an antihypertensive agent, and an antiglaucoma agent. It has been shown to inhibit H2O2-induced cell damage and scavenge O2, potentially offering protective action against UVB-induced damage.
  • Non-selective beta blocker
  • Beta-adrenergic antagonist
  • Inhibits H2O2-induced cell damage
  • Scavenges O2
  • Offers protective action against UVB-induced damage
  • Used as an anti-arrhythmia agent in research
  • Used as an anti-angina agent in research
  • Used as an antihypertensive agent in research
  • Used as an antiglaucoma agent in research
Catalog No. 50-003-84458 Supplier Medchemexpress LLC Supplier No. 25MGHY17495AR
Carteolol hydrochloride is a non-selective beta blocker and beta-adrenergic antagonist. It is used in research as an anti-arrhythmia agent, an anti-angina agent, an antihypertensive agent, and an antiglaucoma agent. It has been shown to inhibit H2O2-induced cell damage and scavenge O2, potentially offering protective action against UVB-induced damage.
  • Non-selective beta blocker
  • Beta-adrenergic antagonist
  • Inhibits H2O2-induced cell damage
  • Scavenges O2
  • Offers protective action against UVB-induced damage
  • Used as an anti-arrhythmia agent in research
  • Used as an anti-angina agent in research
  • Used as an antihypertensive agent in research
  • Used as an antiglaucoma agent in research