Learn More
Medchemexpress LLC Carteolol hydrochloride | 51781-21-6 | 99.6% | 328.8343 | 5 MG
SDP
Catalog No. 5000387064
Encompass_Preferred
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma, as well as an anti-arrhythmia, anti-angina, and antihypertensive agent. It has a protective action against UVB-induced HCEC damage, with its radical scavenging ability potentially being an important basis for this effect. A new alginate formulation of long-acting carteolol 1% given once daily is as effective as standard 1% carteolol given twice daily, with no significant differences in safety.
  • Used to treat glaucoma
  • Functions as an anti-arrhythmia agent
  • Acts as an anti-angina agent
  • Serves as an antihypertensive agent
  • Protective action against UVB-induced HCEC damage
  • Radical scavenging ability
Catalog No. 50-003-87064 Supplier Medchemexpress LLC Supplier No. HY17495AR5MG
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma, as well as an anti-arrhythmia, anti-angina, and antihypertensive agent. It has a protective action against UVB-induced HCEC damage, with its radical scavenging ability potentially being an important basis for this effect. A new alginate formulation of long-acting carteolol 1% given once daily is as effective as standard 1% carteolol given twice daily, with no significant differences in safety.
  • Used to treat glaucoma
  • Functions as an anti-arrhythmia agent
  • Acts as an anti-angina agent
  • Serves as an antihypertensive agent
  • Protective action against UVB-induced HCEC damage
  • Radical scavenging ability