Learn More
Medchemexpress LLC Carteolol hydrochloride | 51781-21-6 | 99.8% | 328.83 | 25 MG
SDP
Catalog No. 5000384332
Encompass_Preferred
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma. For research use only. We do not sell to patients. Carteolol HCl is a beta-adrenergic antagonist used as an anti-arrhythmia agent, an anti-angina agent, an antihypertensive agent, and an antiglaucoma agent.
  • Significantly inhibited H2O2-induced cell damage at 1 mmol/L (P<0.05) and scavenged O2 (EC50 value: 48 mmol/L).
  • Exhibits protective action against UVB-induced HCEC damage, possibly due to its radical scavenging ability.
  • A new alginate formulation of long-acting carteolol 1% given once daily is as effective as standard 1% carteolol given twice daily for glaucoma treatment, with comparable safety.
Catalog No. 50-003-84332 Supplier Medchemexpress LLC Supplier No. 25MGHY17495A
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma. For research use only. We do not sell to patients. Carteolol HCl is a beta-adrenergic antagonist used as an anti-arrhythmia agent, an anti-angina agent, an antihypertensive agent, and an antiglaucoma agent.
  • Significantly inhibited H2O2-induced cell damage at 1 mmol/L (P<0.05) and scavenged O2 (EC50 value: 48 mmol/L).
  • Exhibits protective action against UVB-induced HCEC damage, possibly due to its radical scavenging ability.
  • A new alginate formulation of long-acting carteolol 1% given once daily is as effective as standard 1% carteolol given twice daily for glaucoma treatment, with comparable safety.