Learn More
Medchemexpress LLC Carteolol hydrochloride | 51781-21-6 | 99.96% | 328.83 | 500 MG
SDP
Catalog No. 5000386899
Encompass_Preferred
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma. It is a beta-adrenergic antagonist used as an anti-arrhythmia, anti-angina, antihypertensive, and antiglaucoma agent. It significantly inhibited H2O2-induced cell damage and was able to scavenge O2, showing protective action against UVB-induced HCEC damage.
  • Non-selective beta blocker
  • Treats glaucoma
  • Anti-arrhythmia agent
  • Anti-angina agent
  • Antihypertensive agent
  • Protective action against UVB-induced HCEC damage in vitro
  • Radical scavenging ability (scavenges O2)
  • Long-acting alginate formulation is as effective as standard for glaucoma treatment
Catalog No. 50-003-86899 Supplier Medchemexpress LLC Supplier No. HY17495A500MG
Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma. It is a beta-adrenergic antagonist used as an anti-arrhythmia, anti-angina, antihypertensive, and antiglaucoma agent. It significantly inhibited H2O2-induced cell damage and was able to scavenge O2, showing protective action against UVB-induced HCEC damage.
  • Non-selective beta blocker
  • Treats glaucoma
  • Anti-arrhythmia agent
  • Anti-angina agent
  • Antihypertensive agent
  • Protective action against UVB-induced HCEC damage in vitro
  • Radical scavenging ability (scavenges O2)
  • Long-acting alginate formulation is as effective as standard for glaucoma treatment