Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Cartesian Diver Set
GREENER_CHOICESCI_ED

Catalog No. S160142 Shop All Supertek Scientific Products
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S160142 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160142 Supplier Supertek™ Supplier No. PH10634
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

By using the Supertek™ Cartesian Diver Set, you can demonstrate the transmission of pressure through liquids, the compressibility of gases, and Archimedes' principle.

  • Cartesian diver set

Specifications

Grade 9 to 12
Includes Cartesian diver, glass cylinder, rubber diaphragms, elastic bands, instructions
Class Size 1 Student
Product Type Diver Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.