Learn More
Tosoh Bioscience LLC CARTR. GUARD HOLDER FOR 2MMID
Supplier:  Tosoh Bioscience LLC 0019308
Encompass
Holder, Guard Cartridge; For 2mm I.D. guard cartridges; 2mm I.D. x 1cm Length
Catalog No. 50-851-541