Learn More
Hach Company Hach Cartrical Sensor Cartridge for AN-ISE sc / A-ISE sc / N-ISE sc
Supplier:  Hach Company LZY694
Encompass
Replacement sensor cartridge designed for AN-ISE sc, A-ISE sc, and N-ISE sc probes. The cartridge contains calibrated ion-selective electrodes for ammonium and potassium, a reference system, and a temperature sensor. It is factory calibrated and RFID-enabled to minimize maintenance and allow immediate operation after replacement.
  • Replacement cartridge for ammonium and potassium ion-selective electrode probes.
  • Contains calibrated electrodes, reference system, and temperature sensor in a compact housing.
  • Factory calibrated and RFID-enabled for automatic recognition after replacement.
  • Simple replacement procedure requires unscrewing the old cartridge and plugging in the new one.
  • Typical service life of approximately 6 months.
Catalog No. NC3253178