Learn More
Waters Corp Atlantis T3 VanGuard Cartridge, 100Å, 3 µm, 3.9 mm X 5 mm, 3/pk
Catalog No. NC1602108 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis T3 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. Atlantis T3 analytical columns containing 3 µm sorbent particles.
Catalog No. NC1602108 Supplier Waters Corp Supplier No. 186007676
Atlantis T3 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. Atlantis T3 analytical columns containing 3 µm sorbent particles.