Learn More
Waters Corp Waters Corporation Check Valve Cartridge Kit, Synthetic Ruby, 4 per Package
Catalog No. NC1727901 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Check Valves are made of synthetic ruby and are contained in cartridges that are easy to replace.
  • Cartridges are easy to replace.
  • Made of synthetic ruby.
  • Compatible with Quaternary Gradient Managers and (Q) Binary Gradient Managers.
  • Includes 4 units per package.
  • Designed for HPLC and purification systems.
  • Compatible with SFC Prep 100 System and Auto Purification instruments.
  • Replacement can be done as required.
Catalog No. NC1727901 Supplier Waters Corp Supplier No. 700001493
The Check Valves are made of synthetic ruby and are contained in cartridges that are easy to replace.
  • Cartridges are easy to replace.
  • Made of synthetic ruby.
  • Compatible with Quaternary Gradient Managers and (Q) Binary Gradient Managers.
  • Includes 4 units per package.
  • Designed for HPLC and purification systems.
  • Compatible with SFC Prep 100 System and Auto Purification instruments.
  • Replacement can be done as required.