Learn More
Grainger Honeywell Respirator Cartridge, Ammonia and Methylamine, Green, 2/Pk.
Catalog No. 502789374
Encompass_Preferred
This air-purifying respirator cartridge is designed for protection against ammonia and methylamine vapors. It features a threaded connection type for secure attachment to compatible respirators and is color-coded green for easy identification of the filter type. This cartridge is intended for use in environments where respiratory protection against these specific chemicals is required.
  • Compatible with 5400, 5500, 7600, 7700, RU6500, RU8500, and RU8800 series respirators
  • Provides protection against ammonia and methylamine
  • NIOSH-rated cartridge
  • Threaded connection type
  • Not for use in atmospheres containing less than 19.5 percent oxygen
Catalog No. 50-278-9374 Supplier Grainger Supplier No. 16M240
This air-purifying respirator cartridge is designed for protection against ammonia and methylamine vapors. It features a threaded connection type for secure attachment to compatible respirators and is color-coded green for easy identification of the filter type. This cartridge is intended for use in environments where respiratory protection against these specific chemicals is required.
  • Compatible with 5400, 5500, 7600, 7700, RU6500, RU8500, and RU8800 series respirators
  • Provides protection against ammonia and methylamine
  • NIOSH-rated cartridge
  • Threaded connection type
  • Not for use in atmospheres containing less than 19.5 percent oxygen