Learn More
Waters Corp Waters Cartridge Heater, Replacement Ion Block, MS Systems, Xevo/Synapt G2
Catalog No. 50792971 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge heater serves as a crucial replacement component for the ion block in various mass spectrometry (MS) systems. It is compatible with a wide array of instruments, including models from the Xevo and Synapt G2 series, and is often included in performance maintenance kits or available as a separate orderable part for system upkeep.
  • Replacement ion block cartridge heater.
  • Compatible with Xevo QTOF, Xevo TQ, SQD2, Xevo TQD, Xevo G2, and Synapt G2 instruments.
  • Designed for MS system types.
  • Supplied as 1 unit per package.
  • Replacement as required for maintenance.
  • Included in several performance maintenance kits for Xevo G2 QTof/Tof instruments.
Catalog No. 50-792-971 Supplier Waters Corp Supplier No. 700004332
This cartridge heater serves as a crucial replacement component for the ion block in various mass spectrometry (MS) systems. It is compatible with a wide array of instruments, including models from the Xevo and Synapt G2 series, and is often included in performance maintenance kits or available as a separate orderable part for system upkeep.
  • Replacement ion block cartridge heater.
  • Compatible with Xevo QTOF, Xevo TQ, SQD2, Xevo TQD, Xevo G2, and Synapt G2 instruments.
  • Designed for MS system types.
  • Supplied as 1 unit per package.
  • Replacement as required for maintenance.
  • Included in several performance maintenance kits for Xevo G2 QTof/Tof instruments.