Learn More
Waters Corp Waters Cartridge Heater, For Ceramic Block, For Ion Block, Mass Spectrometry
Catalog No. 50466644 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge heater is designed for use in mass spectrometry (MS) systems, specifically for heating the ion block. It is compatible with a wide range of mass spectrometry instruments, including models from Xevo, Synapt, and SELECT SERIES.
  • Designed for use in mass spectrometry (MS) systems.
  • Provides heating for ion block applications.
  • Compatible with a wide range of laboratory instruments for mass spectrometry.
  • Replacement is scheduled as required.
Catalog No. 50-466-644 Supplier Waters Corp Supplier No. 700005347
This cartridge heater is designed for use in mass spectrometry (MS) systems, specifically for heating the ion block. It is compatible with a wide range of mass spectrometry instruments, including models from Xevo, Synapt, and SELECT SERIES.
  • Designed for use in mass spectrometry (MS) systems.
  • Provides heating for ion block applications.
  • Compatible with a wide range of laboratory instruments for mass spectrometry.
  • Replacement is scheduled as required.