Learn More
IDEX Health & Science Upchurch CARTRIDGE HOLDER BP W/F-300
Catalog No. NC1920557
Encompass_Preferred
Idex Back Pressure Regulators: Cartridge Holders, Description: High pressure; For 1/16 in. OD tubing, For Use With: Back Pressure Regulators
Catalog No. NC1920557 Supplier IDEX Health & Science Upchurch Supplier No. U469
Idex Back Pressure Regulators: Cartridge Holders, Description: High pressure; For 1/16 in. OD tubing, For Use With: Back Pressure Regulators