Learn More
Bio Rad Laboratories CARTRIDGE HOLDER DE-ASHING
Catalog No. 509012171
Encompass
Cartridge Holder, De-Ashing; Bio-Rad; For de-ashing refill cartridges; Holds two 3.0 x 4.6mm cartridges in series
Catalog No. 50-901-2171 Supplier Bio Rad Laboratories Supplier No. 1250139
Cartridge Holder, De-Ashing; Bio-Rad; For de-ashing refill cartridges; Holds two 3.0 x 4.6mm cartridges in series