Learn More
Waters Corp Waters Corporation Cartridge Holder, 10 mm x 10 mm, for Preparative Chromatography
Catalog No. 50782490 Shop All Waters Corporation Products
Encompass_Preferred
The Cartridge Holder, 10 mm x 10 mm, supports various cartridges for preparative chromatography. It easily integrates into existing liquid chromatography systems without external utilities or end-fittings. This holder increases lab efficiency and reduces operating costs.
  • Supports various cartridges for preparative chromatographic needs.
  • Easily incorporated into existing liquid chromatography systems.
  • No end-fittings required for connection or reconnection.
  • Increases lab efficiency and reduces operating costs.
  • Replacement o-rings available for optimized performance and extended lifetime.
  • Manufactured in facilities adhering to cGMP and ISO 9001:2000 standards.
  • Uses ultra-pure reagents; stringent batch testing ensures quality and reproducible performance.
Catalog No. 50-782-490 Supplier Waters Corp Supplier No. 289000779
The Cartridge Holder, 10 mm x 10 mm, supports various cartridges for preparative chromatography. It easily integrates into existing liquid chromatography systems without external utilities or end-fittings. This holder increases lab efficiency and reduces operating costs.
  • Supports various cartridges for preparative chromatographic needs.
  • Easily incorporated into existing liquid chromatography systems.
  • No end-fittings required for connection or reconnection.
  • Increases lab efficiency and reduces operating costs.
  • Replacement o-rings available for optimized performance and extended lifetime.
  • Manufactured in facilities adhering to cGMP and ISO 9001:2000 standards.
  • Uses ultra-pure reagents; stringent batch testing ensures quality and reproducible performance.