Learn More
Waters Corp Waters Cartridge Holder, 19 mm, 10 mm
Catalog No. 50818795 Shop All Waters Corporation Products
Encompass_Preferred
This Cartridge Holder is designed to support and fit cartridges that are 10 mm in length, with a 19 mm diameter. It seamlessly integrates into existing liquid chromatography systems without requiring external utilities or column end-fitting connections, making it suitable for preparative chromatographic applications.
  • Supports cartridges 10 mm in length and 19 mm in diameter.
  • Increases analytical column lifetime.
  • May enhance system loadability.
  • Incorporates into existing liquid chromatography systems without external utilities.
  • Manufactured to cGMP and ISO 9001:2000 industry standards, utilizing ultra-pure reagents.
  • Replacement o-rings are available for optimized performance and extended product lifetime.
  • Increases lab efficiency and reduces overall operating costs.
Catalog No. 50-818-795 Supplier Waters Corp Supplier No. 186000709
This Cartridge Holder is designed to support and fit cartridges that are 10 mm in length, with a 19 mm diameter. It seamlessly integrates into existing liquid chromatography systems without requiring external utilities or column end-fitting connections, making it suitable for preparative chromatographic applications.
  • Supports cartridges 10 mm in length and 19 mm in diameter.
  • Increases analytical column lifetime.
  • May enhance system loadability.
  • Incorporates into existing liquid chromatography systems without external utilities.
  • Manufactured to cGMP and ISO 9001:2000 industry standards, utilizing ultra-pure reagents.
  • Replacement o-rings are available for optimized performance and extended product lifetime.
  • Increases lab efficiency and reduces overall operating costs.