Learn More
Waters Corp Waters Corporation Cartridge Holder, 7.8 mm X 10 mm, for Liquid Chromatography
Catalog No. 50819091 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Cartridge Holder is designed to hold and fit cartridge columns with a length of 7.8 mm x 10 mm. It helps extend the lifetime of analytical columns and can improve system loadability. It requires no external utilities and can be easily incorporated into an existing liquid chromatography system. The columns cannot be joined or reconnected using end fittings.
  • Designed to hold and fit cartridge columns with a length of 7.8 mm x 10 mm.
  • Reduces overall chromatographic performance reduction from common preparative guard columns.
  • May significantly extend the lifetime of analytical columns.
  • May improve system loadability.
  • Requires no external utilities.
  • Easy to incorporate into an existing liquid chromatography system.
  • Manufactured to cGMP and ISO 9001:2000 standards.
  • Produced using ultra-pure reagents.
  • Batch tested for quality.
Catalog No. 50-819-091 Supplier Waters Corp Supplier No. 186000708
This Cartridge Holder is designed to hold and fit cartridge columns with a length of 7.8 mm x 10 mm. It helps extend the lifetime of analytical columns and can improve system loadability. It requires no external utilities and can be easily incorporated into an existing liquid chromatography system. The columns cannot be joined or reconnected using end fittings.
  • Designed to hold and fit cartridge columns with a length of 7.8 mm x 10 mm.
  • Reduces overall chromatographic performance reduction from common preparative guard columns.
  • May significantly extend the lifetime of analytical columns.
  • May improve system loadability.
  • Requires no external utilities.
  • Easy to incorporate into an existing liquid chromatography system.
  • Manufactured to cGMP and ISO 9001:2000 standards.
  • Produced using ultra-pure reagents.
  • Batch tested for quality.