Learn More
Waters Corp Waters Check Valve, i2V, Biocompatible, UPLC System, 1 per Package
Catalog No. 502039534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement biocompatible i2V cartridge is designed for use in various ACQUITY and M-Class Solvent Manager systems, ensuring reliable operation.
  • Replacement cartridge for inlet check valve
  • Biocompatible i2V design
  • Compatible with ACQUITY H-Class Bio, ACQUITY Premier, Arc Premier, and M-Class Solvent Managers
  • For UPLC system type
  • One unit per package
Catalog No. 50-203-9534 Supplier Waters Corp Supplier No. 700005414
This replacement biocompatible i2V cartridge is designed for use in various ACQUITY and M-Class Solvent Manager systems, ensuring reliable operation.
  • Replacement cartridge for inlet check valve
  • Biocompatible i2V design
  • Compatible with ACQUITY H-Class Bio, ACQUITY Premier, Arc Premier, and M-Class Solvent Managers
  • For UPLC system type
  • One unit per package