Learn More
Waters Corp Inject Cartridge, 18 Kpsi, APC SM-FTN
Supplier:  Waters Corp 700009919
Encompass_Preferred
This inject cartridge is a spare part designed for use in injectors operating at up to 18 Kpsi, specifically compatible with APC SM-FTN systems.
  • Designed for injectors
  • Pressure rating of 18 Kpsi
  • Compatible with APC SM-FTN systems
Catalog No. 50-239-0631