Learn More
Evoqua Water Technologies LLC CARTRIDGE LABPURE S5 LC186 EA
Catalog No. NC0802813
Encompass
LABPURE S5 PURIFICATION CARTRIDGE, ELGA LABWATER LC186 SDI FEED Purification cartridge for Ultra systems Model 2 (Blue door units). Compatible with Purelab Ultra Mk1 and Purelab Ultra Mk2 systems.
Catalog No. NC0802813 Supplier Evoqua Water Technologies LLC Supplier No. W2T198828
LABPURE S5 PURIFICATION CARTRIDGE, ELGA LABWATER LC186 SDI FEED Purification cartridge for Ultra systems Model 2 (Blue door units). Compatible with Purelab Ultra Mk1 and Purelab Ultra Mk2 systems.