Learn More
Grainger North By PIP Gas and Vapor Cartridge, Chlorine, Mercury Vapor, Olive, 1 Pair
Catalog No. 502788434
Encompass_Preferred
These gas and vapor cartridges provide respiratory protection against mercury vapor and chlorine. Featuring a threaded design for secure attachment, these cartridges are colored olive for easy identification and are suitable for specialized filtration applications.
  • Designed for protection against mercury vapor and chlorine
  • Olive color coded for identification
  • Threaded connection for reliable fit
Catalog No. 50-278-8434 Supplier Grainger Supplier No. 16M241
These gas and vapor cartridges provide respiratory protection against mercury vapor and chlorine. Featuring a threaded design for secure attachment, these cartridges are colored olive for easy identification and are suitable for specialized filtration applications.
  • Designed for protection against mercury vapor and chlorine
  • Olive color coded for identification
  • Threaded connection for reliable fit