Learn More
Waters Corp Waters Cartridge, Rotary Shear, Stainless Steel
Catalog No. 502039541 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement cartridge valve is made of stainless steel and features a rotary shear design. It is designed for use in UPLC systems.
  • Functions as an injector/valve pod.
  • Used in ACQUITY CM-A and UPC2 CM-30S.
  • Supplied as 1 unit per package.
  • Intended for replacement as required.
  • Part of spare parts kits.
Catalog No. 50-203-9541 Supplier Waters Corp Supplier No. 700005438
This replacement cartridge valve is made of stainless steel and features a rotary shear design. It is designed for use in UPLC systems.
  • Functions as an injector/valve pod.
  • Used in ACQUITY CM-A and UPC2 CM-30S.
  • Supplied as 1 unit per package.
  • Intended for replacement as required.
  • Part of spare parts kits.