Learn More
AVANTOR MASTERFLEX (VWR LLC) Pentair WP1BB20P Fibrous polypropylene string-wound cartridge, 1 m, 20"" length
Catalog No. 50632258
Encompass
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165¿F (74 ¿C).
Catalog No. 50-632-258 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2980237
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165¿F (74 ¿C).